Assay Details
| Gene Symbol: Tpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173082.1 | NP_001166553.1 | 4 - 5 | 500 | 86 | |
| GenBank mRNA | EU330893.1 | - | 4 - 5 | 500 | 86 | |
More Information
Set Membership: |
|

Tpt1
Genomic Map