Assay Details
| Gene Symbol: Nras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172898.1 | NP_001166369.1 | 1 - 2 | 109 | 88 | |
| XM_005007753.3 | XP_005007810.1 | 2 - 3 | 251 | 88 | ||
| GenBank mRNA | X60128.1 | - | 2 - 3 | 179 | 88 | |
More Information
Set Membership: |
|

Nras
Genomic Map