Assay Details
| Gene Symbol: Crygs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173080.1 | NP_001166551.1 | 2 - 3 | 304 | 83 | |
| GenBank mRNA | DQ903939.1 | - | 2 - 3 | 304 | 83 | |
| EB371741.1 | - | 2 - 3 | 289 | 83 | ||
More Information
Set Membership: |
|

Crygs
Genomic Map