Assay Details
| Gene Symbol: Crygc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173093.1 | NP_001166564.1 | 2 - 3 | 281 | 79 | |
| GenBank mRNA | EB371086.1 | - | 2 - 3 | 281 | 79 | |
| EF426306.1 | - | 2 - 3 | 281 | 79 | ||
More Information
Set Membership: |
|

Crygc
Genomic Map