Assay Details
| Gene Symbol: Svp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172730.1 | NP_001166201.1 | 5 - 6 | 1864 | 72 | |
| GenBank mRNA | AF001982.1 | - | 3 - 4 | 1534 | 72 | |
| AF002657.1 | - | 5 - 6 | 1864 | 72 | ||
| AF002658.1 | - | 3 - 4 | 1646 | 72 | ||
| J02968.1 | - | 1 - 2 | 622 | 72 | ||
| M33295.1 | - | 2 - 3 | 1183 | 72 | ||
More Information
Set Membership: |
|

Svp
Genomic Map