Assay Details
| Gene Symbol: Tp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172740.1 | NP_001166211.1 | 9 - 10 | 1223 | 78 | |
| GenBank mRNA | AJ009673.1 | - | 9 - 10 | 1223 | 78 | |
More Information
Set Membership: |
|

Tp53
Genomic Map