Assay Details
Gene Symbol: Htr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172751.1 | NP_001166222.1 | 1 | 944 | 86 | |
GenBank mRNA | AY344643.1 | - | 1 | 944 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
