Assay Details
| Gene Symbol: Cxcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173416.1 | NP_001166887.1 | 1 | 548 | 63 | |
| XM_013160012.2 | XP_013015466.1 | 3 | 734 | 63 | ||
| XM_013160018.2 | XP_013015472.1 | 4 | 777 | 63 | ||
More Information
Set Membership: |
|

Cxcr1
Genomic Map