Assay Details
| Gene Symbol: Gstm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172834.1 | NP_001166305.1 | 3 - 4 | 248 | 61 | |
| GenBank mRNA | AB000488.1 | - | 3 - 4 | 248 | 61 | |
| Gene Symbol: LOC100731896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005007856.3 | XP_005007913.1 | 3 - 4 | 266 | 61 | |
More Information
Set Membership: |
|

Gstm2
Genomic Map