Assay Details
| Gene Symbol: Cd1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172851.1 | NP_001166322.1 | 4 - 5 | 883 | 69 | |
| GenBank mRNA | AF145485.1 | - | 4 - 5 | 883 | 69 | |
More Information
Set Membership: |
|

Cd1b3
Genomic Map