Assay Details
| Gene Symbol: Cd1b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172852.1 | NP_001166323.1 | 2 - 3 | 316 | 72 | |
| GenBank mRNA | AF145486.1 | - | 2 - 3 | 316 | 72 | |
More Information
Set Membership: |
|

Cd1b4
Genomic Map