Assay Details
| Gene Symbol: Cyp3a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173069.1 | NP_001166540.1 | 2056 | 90 | ||
| GenBank mRNA | D28515.1 | - | 2056 | 90 | ||
| Gene Symbol: Ccni | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003477593.4 | XP_003477641.1 | 7 | 1895 | 90 | |
More Information
Set Membership: |
|

Cyp3a17
Genomic Map