Assay Details
| Gene Symbol: Tbxa2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003460823.4 | XP_003460871.1 | 2 - 3 | 1132 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbxa2r| Gene Symbol: Tbxa2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003460823.4 | XP_003460871.1 | 2 - 3 | 1132 | 79 | |
Set Membership: |
|