Assay Details
| Gene Symbol: Prkag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003476061.4 | XP_003476109.2 | 7 - 8 | 484 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkag1| Gene Symbol: Prkag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003476061.4 | XP_003476109.2 | 7 - 8 | 484 | 76 | |
Set Membership: |
|