Assay Details
| Gene Symbol: Prkab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003478346.4 | XP_003478394.1 | 2 - 3 | 281 | 68 | |
| XM_005007214.3 | XP_005007271.1 | 1 - 2 | 481 | 68 | ||
| XM_013142089.2 | XP_012997543.1 | 2 - 3 | 392 | 68 | ||
More Information
Set Membership: |
|

Prkab1
Genomic Map