Assay Details
| Gene Symbol: MRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135747.4 | NP_609591.2 | 11 - 12 | 4601 | 86 | |
| NM_165020.3 | NP_723772.2 | 11 - 12 | 4608 | 86 | ||
| NM_205968.2 | NP_995690.1 | 11 - 12 | 4601 | 86 | ||
| NM_205969.2 | NP_995691.1 | 12 - 13 | 4719 | 86 | ||
| NM_205970.2 | NP_995692.1 | 12 - 13 | 4719 | 86 | ||
| NM_205971.2 | NP_995693.1 | 11 - 12 | 4604 | 86 | ||
| NM_205972.2 | NP_995694.1 | 11 - 12 | 4601 | 86 | ||
| NM_205973.2 | NP_995695.1 | 12 - 13 | 4719 | 86 | ||
| NM_205974.2 | NP_995696.1 | 11 - 12 | 4601 | 86 | ||
| NM_205975.2 | NP_995697.1 | 11 - 12 | 4601 | 86 | ||
| NM_205976.2 | NP_995698.1 | 11 - 12 | 4601 | 86 | ||
| NM_205977.2 | NP_995699.1 | 12 - 13 | 4719 | 86 | ||
| NM_205978.2 | NP_995700.1 | 11 - 12 | 4601 | 86 | ||
| NM_205979.2 | NP_995701.1 | 12 - 13 | 4719 | 86 | ||
| NM_205980.2 | NP_995702.1 | 12 - 13 | 4722 | 86 | ||
| NM_205981.2 | NP_995703.1 | 12 - 13 | 4719 | 86 | ||
| NM_205982.2 | NP_995704.1 | 12 - 13 | 4719 | 86 | ||
| GenBank mRNA | BT150389.2 | - | 11 - 12 | 4580 | 86 | |
More Information
Set Membership: |
|

MRP
Genomic Map