Assay Details
| Gene Symbol: MRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316390.1 | NP_001303319.1 | 7 - 8 | 3031 | 65 | |
| NM_135747.4 | NP_609591.2 | 6 - 7 | 2913 | 65 | ||
| NM_165020.3 | NP_723772.2 | 6 - 7 | 2917 | 65 | ||
| NM_205968.2 | NP_995690.1 | 6 - 7 | 2913 | 65 | ||
| NM_205969.2 | NP_995691.1 | 7 - 8 | 3031 | 65 | ||
| NM_205970.2 | NP_995692.1 | 7 - 8 | 3031 | 65 | ||
| NM_205971.2 | NP_995693.1 | 6 - 7 | 2913 | 65 | ||
| NM_205972.2 | NP_995694.1 | 6 - 7 | 2913 | 65 | ||
| NM_205973.2 | NP_995695.1 | 7 - 8 | 3031 | 65 | ||
| NM_205974.2 | NP_995696.1 | 6 - 7 | 2913 | 65 | ||
| NM_205975.2 | NP_995697.1 | 6 - 7 | 2913 | 65 | ||
| NM_205976.2 | NP_995698.1 | 6 - 7 | 2913 | 65 | ||
| NM_205977.2 | NP_995699.1 | 7 - 8 | 3031 | 65 | ||
| NM_205978.2 | NP_995700.1 | 6 - 7 | 2913 | 65 | ||
| NM_205979.2 | NP_995701.1 | 7 - 8 | 3031 | 65 | ||
| NM_205980.2 | NP_995702.1 | 7 - 8 | 3031 | 65 | ||
| NM_205981.2 | NP_995703.1 | 7 - 8 | 3031 | 65 | ||
| NM_205982.2 | NP_995704.1 | 7 - 8 | 3031 | 65 | ||
| GenBank mRNA | BT150389.2 | - | 6 - 7 | 2892 | 65 | |
More Information
Set Membership: |
|

MRP
Genomic Map