Assay Details
| Gene Symbol: ACXC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038813.2 | NP_001033902.1 | 9 - 10 | 2812 | 85 | |
| NM_135749.2 | NP_609593.1 | 10 - 11 | 2753 | 85 | ||
| GenBank mRNA | AF177930.1 | - | 10 - 11 | 2754 | 85 | |
| BT021449.1 | - | 10 - 11 | 2740 | 85 | ||
More Information
Set Membership: |
|

ACXC
Genomic Map