Assay Details
| Gene Symbol: Sfmbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144349.2 | NP_001137821.1 | 2 - 3 | 459 | 79 | |
| NM_001273503.1 | NP_001260432.1 | 2 - 3 | 447 | 79 | ||
| NM_135762.3 | NP_609606.2 | 2 - 3 | 459 | 79 | ||
| GenBank mRNA | BT006011.1 | - | 2 - 3 | 457 | 79 | |
More Information
Set Membership: |
|

Sfmbt
Genomic Map