Assay Details
| Gene Symbol: Sfmbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144349.2 | NP_001137821.1 | 5 - 6 | 2841 | 109 | |
| NM_001273503.1 | NP_001260432.1 | 5 - 6 | 2829 | 109 | ||
| NM_135762.3 | NP_609606.2 | 5 - 6 | 2841 | 109 | ||
| NM_165026.2 | NP_723786.1 | 4 - 5 | 2003 | 109 | ||
| GenBank mRNA | BT006011.1 | - | 5 - 6 | 2839 | 109 | |
More Information
Set Membership: |
|

Sfmbt
Genomic Map