Assay Details
| Gene Symbol: Sfmbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273503.1 | NP_001260432.1 | 7 - 8 | 3633 | 65 | |
| NM_135762.3 | NP_609606.2 | 7 - 8 | 3645 | 65 | ||
| NM_165026.2 | NP_723786.1 | 6 - 7 | 2807 | 65 | ||
| GenBank mRNA | BT006011.1 | - | 7 - 8 | 3643 | 65 | |
More Information
Set Membership: |
|

Sfmbt
Genomic Map