Assay Details
| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135765.4 | NP_609609.1 | 1 - 2 | 219 | 67 | |
| GenBank mRNA | AY089373.1 | - | 1 - 2 | 221 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsph1| Gene Symbol: Rsph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135765.4 | NP_609609.1 | 1 - 2 | 219 | 67 | |
| GenBank mRNA | AY089373.1 | - | 1 - 2 | 221 | 67 | |
Set Membership: |
|