Assay Details
| Gene Symbol: CG44085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103686.3 | NP_001097156.1 | 2 - 3 | 1359 | 60 | |
| NM_001103687.3 | NP_001097157.1 | 3 - 4 | 1036 | 60 | ||
| NM_001169497.1 | NP_001162968.1 | 3 - 4 | 1415 | 60 | ||
| NM_001169498.1 | NP_001162969.1 | 3 - 4 | 1180 | 60 | ||
| NM_001169499.1 | NP_001162970.1 | 3 - 4 | 1172 | 60 | ||
| NM_001259091.2 | NP_001246020.1 | 2 - 3 | 1172 | 60 | ||
| NM_135785.3 | NP_609629.1 | 3 - 4 | 1036 | 60 | ||
| NM_135786.2 | NP_609630.2 | 2 - 3 | 844 | 60 | ||
| NM_165034.3 | NP_723801.2 | 2 - 3 | 1172 | 60 | ||
| NM_165035.4 | NP_723802.2 | 3 - 4 | 1036 | 60 | ||
| NM_165037.3 | NP_723804.1 | 4 - 5 | 1414 | 60 | ||
| GenBank mRNA | BT083391.1 | - | 3 - 4 | 1406 | 60 | |
More Information
Set Membership: |
|

CG44085
Genomic Map