Assay Details
| Gene Symbol: CG44085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103687.3 | NP_001097157.1 | 6 - 7 | 2421 | 86 | |
| NM_135785.3 | NP_609629.1 | 6 - 7 | 2421 | 86 | ||
| NM_165037.3 | NP_723804.1 | 7 - 8 | 2799 | 86 | ||
More Information
Set Membership: |
|

CG44085
Genomic Map