Assay Details
| Gene Symbol: CG44085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103686.3 | NP_001097156.1 | 7 - 8 | 3178 | 54 | |
| NM_001103687.3 | NP_001097157.1 | 9 - 10 | 2975 | 54 | ||
| NM_001169498.1 | NP_001162969.1 | 8 - 9 | 2999 | 54 | ||
| NM_001169499.1 | NP_001162970.1 | 8 - 9 | 2991 | 54 | ||
| NM_001259090.1 | NP_001246019.1 | 7 - 8 | 2629 | 54 | ||
| NM_001273511.1 | NP_001260440.1 | 7 - 8 | 2646 | 54 | ||
| NM_135785.3 | NP_609629.1 | 8 - 9 | 2903 | 54 | ||
| GenBank mRNA | BT088972.1 | - | 7 - 8 | 2629 | 54 | |
More Information
Set Membership: |
|

CG44085
Genomic Map