Assay Details
| Gene Symbol: CG44085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103686.3 | NP_001097156.1 | 8 - 9 | 3223 | 66 | |
| NM_001103687.3 | NP_001097157.1 | 10 - 11 | 3020 | 66 | ||
| NM_001169497.1 | NP_001162968.1 | 7 - 8 | 2841 | 66 | ||
| NM_001169498.1 | NP_001162969.1 | 9 - 10 | 3044 | 66 | ||
| NM_001169499.1 | NP_001162970.1 | 9 - 10 | 3036 | 66 | ||
| NM_001259090.1 | NP_001246019.1 | 8 - 9 | 2674 | 66 | ||
| NM_001259091.2 | NP_001246020.1 | 9 - 10 | 3120 | 66 | ||
| NM_001273511.1 | NP_001260440.1 | 8 - 9 | 2691 | 66 | ||
| NM_135785.3 | NP_609629.1 | 9 - 10 | 2948 | 66 | ||
| NM_135786.2 | NP_609630.2 | 6 - 7 | 2270 | 66 | ||
| NM_165034.3 | NP_723801.2 | 6 - 7 | 2598 | 66 | ||
| NM_165035.4 | NP_723802.2 | 7 - 8 | 2462 | 66 | ||
| NM_165037.3 | NP_723804.1 | 9 - 10 | 3048 | 66 | ||
| GenBank mRNA | BT083391.1 | - | 7 - 8 | 2832 | 66 | |
| BT088972.1 | - | 8 - 9 | 2674 | 66 | ||
More Information
Set Membership: |
|

CG44085
Genomic Map