Assay Details
| Gene Symbol: CG44085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273511.1 | NP_001260440.1 | 10 | 7346 | 62 | |
| NM_135786.2 | NP_609630.2 | 8 | 6925 | 62 | ||
More Information
Set Membership: |
|

CG44085
Genomic Map