Assay Details
| Gene Symbol: Drep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135787.4 | NP_609631.1 | 2 - 3 | 420 | 62 | |
| GenBank mRNA | AF149797.2 | - | 2 - 3 | 367 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Drep4| Gene Symbol: Drep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135787.4 | NP_609631.1 | 2 - 3 | 420 | 62 | |
| GenBank mRNA | AF149797.2 | - | 2 - 3 | 367 | 62 | |
Set Membership: |
|