Assay Details
| Gene Symbol: CG9395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169502.2 | NP_001162973.2 | 2 - 3 | 680 | 82 | |
| NM_135795.3 | NP_609639.2 | 1 - 2 | 973 | 82 | ||
More Information
Set Membership: |
|

CG9395
Genomic Map