Assay Details
| Gene Symbol: CG9395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169502.2 | NP_001162973.2 | 3 - 4 | 802 | 67 | |
| NM_135795.3 | NP_609639.2 | 2 - 3 | 1095 | 67 | ||
More Information
Set Membership: |
|

CG9395
Genomic Map