Assay Details
| Gene Symbol: CG9395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169502.2 | NP_001162973.2 | 4 - 5 | 885 | 62 | |
| NM_135795.3 | NP_609639.2 | 3 - 4 | 1178 | 62 | ||
More Information
Set Membership: |
|

CG9395
Genomic Map