Assay Details
| Gene Symbol: CG9395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169502.2 | NP_001162973.2 | 5 - 6 | 1294 | 68 | |
| NM_135795.3 | NP_609639.2 | 4 - 5 | 1587 | 68 | ||
More Information
Set Membership: |
|

CG9395
Genomic Map