Assay Details
| Gene Symbol: ics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298973.1 | NP_001285902.1 | 2 - 3 | 779 | 63 | |
| NM_001298974.1 | NP_001285903.1 | 3 - 4 | 1360 | 63 | ||
| NM_135821.5 | NP_609665.2 | 4 - 5 | 1051 | 63 | ||
| GenBank mRNA | BT031285.1 | - | 2 - 3 | 779 | 63 | |
| BT088923.1 | - | 4 - 5 | 1017 | 63 | ||
More Information
Set Membership: |
|

ics
Genomic Map