Assay Details
| Gene Symbol: Sec71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135831.3 | NP_609675.2 | 2 - 3 | 5155 | 63 | |
| GenBank mRNA | AY122171.1 | - | 2 - 3 | 5126 | 63 | |
More Information
Set Membership: |
|

Sec71
Genomic Map