Assay Details
| Gene Symbol: Adat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273521.1 | NP_001260450.1 | 1 - 2 | 893 | 154 | |
| NM_135832.3 | NP_609676.1 | 1 - 2 | 893 | 154 | ||
| GenBank mRNA | AF192530.1 | - | 1 - 2 | 891 | 154 | |
More Information
Set Membership: |
|

Adat1
Genomic Map