Assay Details
| Gene Symbol: Smg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298985.1 | NP_001285914.1 | 3 - 4 | 1897 | 60 | |
| NM_135841.5 | NP_609685.1 | 3 - 4 | 2057 | 60 | ||
| NM_165069.2 | NP_723851.1 | 3 - 4 | 2063 | 60 | ||
| GenBank mRNA | AJ556817.1 | - | 2 - 3 | 1782 | 60 | |
| BT031137.1 | - | 3 - 4 | 1835 | 60 | ||
More Information
Set Membership: |
|

Smg5
Genomic Map