Assay Details
| Gene Symbol: NimA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259097.1 | NP_001246026.1 | 2 - 3 | 796 | 97 | |
| NM_001298989.1 | NP_001285918.1 | 2 - 3 | 550 | 97 | ||
| NM_135847.6 | NP_609691.6 | 2 - 3 | 796 | 97 | ||
| GenBank mRNA | BT100252.1 | - | 2 - 3 | 796 | 97 | |
More Information
Set Membership: |
|

NimA
Genomic Map