Assay Details
| Gene Symbol: CG46301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347776.1 | NP_001334738.1 | 2 - 3 | 803 | 71 | |
| NM_001347777.1 | NP_001334737.1 | 2 - 3 | 803 | 71 | ||
| NM_001347778.1 | NP_001334739.1 | 2 - 3 | 803 | 71 | ||
More Information
Set Membership: |
|

CG46301
Genomic Map