Assay Details
| Gene Symbol: CG46301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347777.1 | NP_001334737.1 | 5 - 6 | 2795 | 62 | |
| NM_135859.4 | NP_609703.2 | 2 - 3 | 1211 | 62 | ||
More Information
Set Membership: |
|

CG46301
Genomic Map