Assay Details
| Gene Symbol: CG46301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347777.1 | NP_001334737.1 | 7 - 8 | 3697 | 134 | |
| NM_135859.4 | NP_609703.2 | 4 - 5 | 2113 | 134 | ||
More Information
Set Membership: |
|

CG46301
Genomic Map