Assay Details
| Gene Symbol: CSN1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135876.2 | NP_609720.1 | 1 | 1049 | 144 | |
| GenBank mRNA | BT022388.1 | - | 1 | 1049 | 144 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSN1a| Gene Symbol: CSN1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135876.2 | NP_609720.1 | 1 | 1049 | 144 | |
| GenBank mRNA | BT022388.1 | - | 1 | 1049 | 144 | |
Set Membership: |
|