Assay Details
| Gene Symbol: Uxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273535.1 | NP_001260464.1 | 1 - 2 | 112 | 69 | |
| NM_135900.4 | NP_609744.2 | 1 - 2 | 112 | 69 | ||
| GenBank mRNA | BT023678.1 | - | 1 - 2 | 84 | 69 | |
| KX531617.1 | - | 1 - 2 | 42 | 69 | ||
More Information
Set Membership: |
|

Uxt
Genomic Map