Assay Details
| Gene Symbol: CG4935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135938.3 | NP_609782.2 | 2 - 3 | 256 | 95 | |
| GenBank mRNA | AY113244.1 | - | 2 - 3 | 274 | 95 | |
More Information
Set Membership: |
|

CG4935
Genomic Map