Assay Details
| Gene Symbol: Prosbeta4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273582.1 | NP_001260511.1 | 2 - 3 | 415 | 89 | |
| NM_135960.4 | NP_609804.1 | 2 - 3 | 415 | 89 | ||
| GenBank mRNA | AY113632.1 | - | 2 - 3 | 412 | 89 | |
More Information
Set Membership: |
|

Prosbeta4
Genomic Map