Assay Details
| Gene Symbol: BuGZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103697.3 | NP_001097167.1 | 3 - 4 | 736 | 66 | |
| NM_001259117.3 | NP_001246046.1 | 3 - 4 | 736 | 66 | ||
| NM_001259118.3 | NP_001246047.1 | 3 - 4 | 736 | 66 | ||
| NM_001273583.2 | NP_001260512.1 | 3 - 4 | 730 | 66 | ||
| NM_001273584.2 | NP_001260513.1 | 3 - 4 | 736 | 66 | ||
| NM_001299057.1 | NP_001285986.1 | 3 - 4 | 736 | 66 | ||
| NM_135964.3 | NP_609808.1 | 3 - 4 | 736 | 66 | ||
| GenBank mRNA | AY051534.1 | - | 3 - 4 | 737 | 66 | |
| Gene Symbol: RpL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT030448.1 | - | 1596 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT030448.1 transcripts.
Set Membership: |
|

BuGZ
Genomic Map