Assay Details
| Gene Symbol: mdy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135969.4 | NP_609813.1 | 2 - 3 | 495 | 103 | |
| NM_165194.3 | NP_724016.1 | 2 - 3 | 422 | 103 | ||
| NM_165195.3 | NP_724017.1 | 2 - 3 | 742 | 103 | ||
| NM_206002.2 | NP_995724.2 | 2 - 3 | 714 | 103 | ||
| GenBank mRNA | AY051835.1 | - | 2 - 3 | 461 | 103 | |
More Information
Set Membership: |
|

mdy
Genomic Map