Assay Details
| Gene Symbol: mdy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135969.4 | NP_609813.1 | 5 - 6 | 773 | 97 | |
| NM_165194.3 | NP_724016.1 | 5 - 6 | 700 | 97 | ||
| NM_165195.3 | NP_724017.1 | 5 - 6 | 1020 | 97 | ||
| NM_206002.2 | NP_995724.2 | 5 - 6 | 992 | 97 | ||
| GenBank mRNA | AY051835.1 | - | 5 - 6 | 739 | 97 | |
More Information
Set Membership: |
|

mdy
Genomic Map