Assay Details
| Gene Symbol: mdy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135969.4 | NP_609813.1 | 8 - 9 | 1340 | 93 | |
| NM_165194.3 | NP_724016.1 | 8 - 9 | 1267 | 93 | ||
| NM_165195.3 | NP_724017.1 | 8 - 9 | 1587 | 93 | ||
| NM_206002.2 | NP_995724.2 | 8 - 9 | 1559 | 93 | ||
| GenBank mRNA | AY051835.1 | - | 8 - 9 | 1306 | 93 | |
More Information
Set Membership: |
|

mdy
Genomic Map