Assay Details
| Gene Symbol: bur | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299191.1 | NP_001286120.1 | 4 - 5 | 1071 | 70 | |
| NM_138759.4 | NP_620114.3 | 4 - 5 | 1071 | 70 | ||
| GenBank mRNA | AY118572.1 | - | 4 - 5 | 1048 | 70 | |
More Information
Set Membership: |
|

bur
Genomic Map