Assay Details
| Gene Symbol: CG9389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141038.3 | NP_649295.1 | 1 - 2 | 1165 | 83 | |
| GenBank mRNA | AY060713.1 | - | 1 - 2 | 1116 | 83 | |
More Information
Set Membership: |
|

CG9389
Genomic Map